Learn More
Thermo Scientific™ Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar w/2D Barcode
Shop All Thermo Scientific ProductsDescription
Experience next-level cleanroom confidence with these plates, complete with VHP indicator, lecithin, polysorbate 80, sodium thiosulphate, L-histidine and 2D barcode.
Experience next-level cleanroom confidence with Thermo Scientific™ Triple Wrap Sterile Pack Tryptone Soya Agar Plates with VHP Indicator and 2D Barcode.
Achieve confidence in your results and ensure optimum media growth using plates with a vapor-phase hydrogen peroxide (VHP) exposure indicator, designed to confirm the integrity of the wrapping material and seal during VHP exposure.
Three protective layers reduce risk of contamination:
- Primary film with moisture control patch
- Secondary film VHP barrier with immobilized desiccant sachet
- Tertiary film dust cover
Conveniently sold in packs of 100 which are sealed for security
The incorporation of novel elements enhance moisture control by permitting the passage and absorption of excess moisture.
Using plates with an improved shelf life and room temperature storage offers a cost-effective solution, minimizing new batch validation and refrigeration costs.
The terms "settle plates" and "settling plates" in the context of environmental monitoring are interchangeable and both refer to passive sampling used to monitor airborne microbial contamination.
Not all products are available for sale in all territories. Please inquire.
Specifications
Specifications
| Color | Ivory, transparent |
| Format | 90 mm monoplate |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar |
| Fill Weight | 30 g |
| Includes | VHP Indicator, Neutralisers |
| Packaging Type | Triple Wrapped - Irradiated |
| Quantity | 100/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.