missing translation for 'onlineSavingsMsg'
Learn More

Tripterin, MedChemExpress

Product Code. 16476096
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. Quantity unitSize
16476096 100 mg 100mg
16411707 50 mg 50mg
16405538 10 mg 10mg
3 options
This item is not returnable. View return policy

Product Code. 16476096

Brand: Medchem Express HY13067

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tripterin (Celastrol) is a proteasome inhibitor which potently and preferentially inhibits the chymotrypsin-like activity of a purified 20S proteasome with IC50 of 2.5 μM.

Tripterin (Celastrol) is a proteasome inhibitor which potently and preferentially inhibits the chymotrypsin-like activity of a purified 20S proteasome with IC50 of 2.5 μM.

Specifications

Chemical Name or Material Tripterin
Physical Form Solid
CAS 34157-83-0
Color Orange
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C29H38O4
For Use With (Application) COVID-19-immunoregulation
Synonym Celastrol
Solubility Information DMSO : 33.33 mg/mL (73.97 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES OC1=C(C2=CC=C3[C@](C)([C@]4(CC[C@]3(C2=CC1=O)C)C)CC[C@@]5(C)CC[C@@](C(O)=O)(C[C@]54[H])C)C
Molecular Weight (g/mol) 450.61
Formula Weight 450.61
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.