missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TRIS, 1.0M buffer soln., pH 8.8

Product Code. 15453219
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453219 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15453219 Supplier Thermo Scientific Chemicals Supplier No. J60452.K2
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

TRIS, 1.0M buffer is used as buffers for stabilizing and storing nucleic acids and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TRIS, 1.0M buffer is used as buffers for stabilizing and storing nucleic acids and proteins.

Solubility
Soluble in water and organic solvents.

Notes
Store in cool place. Keep container tightly close
TRUSTED_SUSTAINABILITY

Specifications

CAS 77-86-1
Concentration 1.0 M
pH 8.8
Physical Form Liquid
Quantity 1 L
Molecular Formula C4H11NO3
Odor Odorless
MDL Number MFCD00004679
Solubility Information Soluble in water and organic solvents.
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
SMILES C(C(CO)(CO)N)O
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular Weight (g/mol) 121.136
PubChem CID 6503
ChEBI CHEBI:9754
Chemical Name or Material TRIS buffer soln.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.