missing translation for 'onlineSavingsMsg'
Learn More

Tris[3,5-bis(trifluoromethyl)phenyl]phosphine, 94%

Product Code. 11397056
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397056 1 g 1g
11307066 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397056 Supplier Thermo Scientific Alfa Aesar Supplier No. L06941.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 175136-62-6 | C24H9F18P | 670.281 g/mol

Tris[3,5-bis(trifluoromethyl)phenyl]phosphine is used as ligands which is useful as catalysts in various reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine is used as ligands which is useful as catalysts in various reactions.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175136-62-6
Molecular Formula C24H9F18P
Molecular Weight (g/mol) 670.281
MDL Number MFCD00042428
InChI Key ITJHLZVYLDBFOJ-UHFFFAOYSA-N
Synonym tris 3,5-bis trifluoromethyl phenyl phosphine, tris 3,5-bis trifluoromethyl phenyl phosphane, acmc-20ajat, tri 3,5-di trifluoromethyl phenyl phosphine, tris 3,5-bis-trifluoromethylphenyl phosphine, phosphine,tris 3,5-bis trifluoromethyl phenyl, tris 3,5-bis trifluoromethyl phenyl phosphine gc , yellow-brown
PubChem CID 2778032
IUPAC Name tris[3,5-bis(trifluoromethyl)phenyl]phosphane
SMILES C1=C(C=C(C=C1C(F)(F)F)P(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)C3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 98°C to 102°C
Boiling Point 128°C to 130°C (0.5 mmHg)
Quantity 1 g
Beilstein 6556841
Solubility Information Insoluble in water.
Formula Weight 670.28
Percent Purity 94%
Chemical Name or Material Tris[3,5-bis(trifluoromethyl)phenyl]phosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.