missing translation for 'onlineSavingsMsg'
Learn More

Tris(3-fluorophenyl)phosphine, 97%

Product Code. 11402078
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11402078

Brand: Thermo Scientific Alfa Aesar A15117.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 23039-94-3 | C18H12F3P | 316.263 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23039-94-3
Molecular Formula C18H12F3P
Molecular Weight (g/mol) 316.263
MDL Number MFCD00015722
InChI Key CUTRINLXFPIWQB-UHFFFAOYSA-N
Synonym tris 3-fluorophenyl phosphine, tris 3-fluorophenyl phosphane, tri 3-fluorophenyl phosphine, phosphine,tris 3-fluorophenyl, acmc-1cqwl, tri m-fluorophenyl phosphine, tri-3-fluorophenyl-phosphine
PubChem CID 140988
IUPAC Name tris(3-fluorophenyl)phosphane
SMILES C1=CC(=CC(=C1)P(C2=CC=CC(=C2)F)C3=CC=CC(=C3)F)F

Specifications

Melting Point 60°C to 63°C
Quantity 1 g
Beilstein 2946854
Formula Weight 316.27
Percent Purity 97%
Chemical Name or Material Tris(3-fluorophenyl)phosphine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.