missing translation for 'onlineSavingsMsg'
Learn More

Tris(4-fluorophenyl)phosphine, 98+%

Product Code. 10115470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10115470 1 g Glass bottle 1g
10269263 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10115470 Supplier Thermo Scientific Acros Supplier No. 306340010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18437-78-0 | C18H12F3P | 316.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18437-78-0
Molecular Formula C18H12F3P
Molecular Weight (g/mol) 316.26
MDL Number MFCD00013553
InChI Key GEPJPYNDFSOARB-UHFFFAOYSA-N
Synonym tris 4-fluorophenyl phosphine, phosphine, tris 4-fluorophenyl, tris p-fluorophenyl phosphine, tri 4-fluorophenyl phosphine, tris 4-fluorophenyl phosphane, phosphine, tris p-fluorophenyl, zlchem 700, pubchem6418, acmc-1c3oi
PubChem CID 140387
IUPAC Name tris(4-fluorophenyl)phosphane
SMILES FC1=CC=C(C=C1)P(C1=CC=C(F)C=C1)C1=CC=C(F)C=C1

Specifications

Melting Point 79.0°C to 82.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula (FC6H4)3P
Quantity 1 g
Formula Weight 316.27
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material Tris(4-fluorophenyl)phosphine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.