missing translation for 'onlineSavingsMsg'
Learn More

Tris(4-methoxyphenyl)phosphine, 95%

Product Code. 10442815 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10442815 1 g Glass bottle 1g
10435471 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10442815 Supplier Thermo Scientific Acros Supplier No. 422240010
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 855-38-9 | C21H21O3P | 352.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 855-38-9
Molecular Formula C21H21O3P
Molecular Weight (g/mol) 352.36
MDL Number MFCD00014896
InChI Key UYUUAUOYLFIRJG-UHFFFAOYSA-N
Synonym tris 4-methoxyphenyl phosphine, trianisylphosphine, tris p-anisyl phosphine, phosphine, tris 4-methoxyphenyl, tri-p-anisylphosphine, tris p-methoxyphenyl phosphine, tris 4-methoxyphenyl phosphane, phosphine, tris p-methoxyphenyl, tri 4-methoxyphenyl phosphine, tri p-methoxyphenyl phosphine
PubChem CID 70071
IUPAC Name tris(4-methoxyphenyl)phosphane
SMILES COC1=CC=C(C=C1)P(C2=CC=C(C=C2)OC)C3=CC=C(C=C3)OC

Specifications

Melting Point 131.0°C to 134.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in methanol
Formula Weight 352.36
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Tris(4-methoxyphenyl)phosphine, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.