missing translation for 'onlineSavingsMsg'
Learn More

Tris(4-trifluoromethylphenyl)phosphine, 99%

Product Code. 10723213 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10723213 1 g Glass bottle 1g
10005873 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10723213 Supplier Thermo Scientific Acros Supplier No. 355340010
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 13406-29-6 | C21H12F9P | 466.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13406-29-6
Molecular Formula C21H12F9P
Molecular Weight (g/mol) 466.29
MDL Number MFCD00058883
InChI Key PXYCJKZSCDFXLR-UHFFFAOYSA-N
Synonym tris 4-trifluoromethylphenyl phosphine, tris 4-trifluoromethyl phenyl phosphine, tris 4-trifluoromethyl phenyl phosphane, phosphine, tris 4-trifluoromethyl phenyl, tris p-trifluoromethylphenyl phosphine, phosphine, tris .alpha.,.alpha.,.alpha.-trifluoro-p-tolyl, tris p-trifluoromethylphenyl phosphine,min, acmc-20ajbq
PubChem CID 139448
IUPAC Name tris[4-(trifluoromethyl)phenyl]phosphane
SMILES C1=CC(=CC=C1C(F)(F)F)P(C2=CC=C(C=C2)C(F)(F)F)C3=CC=C(C=C3)C(F)(F)F

Specifications

Melting Point 72.0°C to 75.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 466.29
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tris(4-trifluoromethylphenyl)phosphine
compliance-icons
Product Identifier
  • Tris(4-trifluoromethylphenyl)phosphine
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H413-May cause long lasting harmful effects to aquatic life.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.