missing translation for 'onlineSavingsMsg'
Learn More

TAE Buffer, Tris-Acetate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents

Product Code. 10628403
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Poly CUBE
Unit Size:
1 L
20 L
4 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10628403 500 mL Poly Bottle 500mL
10041223 1 L Nalgene Poly Bottle 1 L
10775494 4 L Poly CUBE 4 L
10637633 20 L Poly CUBE 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10628403 Supplier Fisher BioReagents Supplier No. BP1335500
Spotlight Offers
Use promo code 28036
View all promotions
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
This item is not returnable. View return policy
  • Save time and simplify your buffer preparation step by using Fisher BioReagents 10X TAE buffer solution - simply dilute as needed
  • 10X solution contains 0.4M tris-acetate and 0.01M EDTA
  • To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TAE Solution with 9 volumes of ultrapure water, such as BP2819
  • Tested for the absence of DNase, RNase, and protease
  • Filtered, autoclaved, and functionally tested for use in electrophoresis applications
  • TAE is a commonly used buffer for electrophoresis of nucleic acids in agarose gels
  • Intended for laboratory research use only
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris-Acetate-EDTA
Name Note 10X Solution
CAS 64-19-7, 7732-18-5, 60-00-4, 77-86-1
Physical Form Liquid
Quantity 500 mL
DNase DNase free
Filtered Through Filtered through a 0.2-micron filter.
Purity Grade Notes DNase-, RNase- and Protease-Free
Packaging Poly Bottle
Protease Protease free
Synonym TAE
Grade Electrophoresis
Concentration or Composition (by Analyte or Components) Tris (<5.0%), Acetic Acid (1.0%), and EDTA (<1%).
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.