missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAE Buffer, Tris-Acetate-EDTA, 25X Powder, Electrophoresis, Fisher BioReagents
Description
- Save time and simplify your buffer preparation step by using Fisher BioReagents 25X TAE Powder -- simply add pure water!
- Each foil pack contains the correct amount of pre-weighed powder to make 1L of 25X TAE buffer containing 1M tris-acetate and 0.025M EDTA
- Tested for the absence of DNase, RNase, and protease
- TAE is a commonly used buffer for electrophoresis of nucleic acids in agarose and polyacrylamide gels
Specifications
Specifications
| Chemical Name or Material | Tris-Acetate-EDTA |
| Name Note | 25X Powder |
| CAS | 77-86-1, 6850-28-8, 6381-92-6, 10378-23-1 |
| Color | White |
| pH | 8.3 |
| Physical Form | Solid |
| Quantity | 1 Pk. |
| DNase | DNase free |
| Packaging | Foil Pack |
| Protease | Protease free |
| Show More |
Safety and Handling
Product Identifier
- Tris Acetate EDTA 25X Powder
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?