missing translation for 'onlineSavingsMsg'
Learn More

Tris Base (White Crystals or Crystalline Powder/Molecular Biology), Fisher BioReagents™

Product Code. 10724344
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
10 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Unit Size:
10kg
1kg
25kg
500g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10724344 5 kg Poly Pail 5kg
10103203 500 g Poly Bottle 500g
10376743 1 kg Poly Bottle 1kg
10667243 10 kg Poly Pail 10kg
10336793 25 kg Fiber Drum 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10724344 Supplier Fisher BioReagents Supplier No. BP1525
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

DNase-, RNase- and Protease-Free. | CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

DNase-, RNase- and Protease-Free.

Tris Base is a buffer component in molecular biology, tissue culture, and electrophoresis procedures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym Trimethylol Aminomethane, Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Absorbance 0.05 AU max. at 280nm
Melting Point 168.5°C
Boiling Point 219°C
Color White
pH 10.4
Physical Form Crystalline Powder or Crystals
Quantity 5 kg
DNase DNase free
Formulation 2-Amino-2-(Hydroxymethyl)-1,3-Propanediol, Tris (hydroxymethyl) Aminomethane, Tris Base, THAM, Trimethylol Aminomethane, Trometamol
Assay Percent Range ≥99.8 %
Identification Pass Test
Linear Formula H2NC(CH2OH)3
Packaging Poly Pail
Protease Protease free
Merck Index 15, 9951
Formula Weight 121.14
Percent Purity ≥99.8%
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material Tris Base
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.