missing translation for 'onlineSavingsMsg'
Learn More

TBE Buffer, 10X Powder, Electrophoresis, Fisher BioReagents

Product Code. 10448543
Change view
Click to view available options
Quantity:
1 Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10448543 1 Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10448543 Supplier Fisher BioReagents Supplier No. BP13341
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Each pack contains the required amount of premixed powder to make 1L of 10X TBE buffer stock solution that can be diluted to a 1X solution for use in electrophoresis of nucleic acids in polyacrylamide gels

  • DNase, RNase, and protease free powder that is pre-mixed in the correct ratios for your convenience
  • Adequate amount of powder to make 1L of 10X TBE buffer
  • Eliminates the need to separately weigh TBE buffer ingredients - simply dissolve powder in pure water
  • TBE buffer is commonly used for DNA and RNA polyacrylamide gel electrophoresis (PAGE)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris-Borate-EDTA
Name Note 10X Powder
CAS 77-86-1, 10043-35-3, 60-00-4
Color White
Physical Form Solid
Quantity 1 Pk.
DNase DNase free
Packaging Foil Pack
Protease Protease free
Synonym TBE
Grade Electrophoresis
Concentration or Composition (by Analyte or Components) Tris (63.91%), Boric Acid (32.54%), and EDTA (3.55%).
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tris borate EDTA 10X powder
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.