missing translation for 'onlineSavingsMsg'
Learn More

TBE Buffer, Tris-Borate-EDTA, 1X Solution, Electrophoresis, Fisher BioReagents

Product Code. 10754914
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Nalgene Poly Bottle
Poly CUBE
Unit Size:
1 L
20 L
4 L
This item is not returnable. View return policy

Product Code. 10754914

Brand: Fisher BioReagents undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
  • Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X TBE buffer solution
  • Eliminates the hassle of dilution or waiting for powder to dissolve
  • 1X solution contains 0.089M tris, 0.089M boric acid, and 0.002M EDTA
  • Tested for the absence of DNase, RNase, and protease
  • Filtered, autoclaved, and functionally tested for use in electrophoresis applications
  • TBE is a commonly used buffer for electrophoresis of nucleic acids in agarose and polyacrylamide gels
  • Intended for laboratory research use only
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris-Borate-EDTA
Name Note 1X Solution
CAS 77-86-1, 6381-92-6, 10043-35-3, 7732-18-5
Color Colorless
pH 8.2 to 8.4
Physical Form Liquid
Quantity 1 L
DNase DNase free
Filtered Through Filtered through a 0.2-micron filter.
Packaging Nalgene Poly Bottle
Protease Protease free
Synonym TBE
Grade Electrophoresis
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.