missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TRIS-buffered saline (TBS, 10X), with 1% Tween 20

Product Code. 15475249
Click to view available options
Quantity:
500 mL
1 L
2 L
Unit Size:
1 L
2 L
500mL
This item is not returnable. View return policy

Product Code. 15475249

Brand: Thermo Scientific Chemicals J62955.K3

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

TRIS-buffered saline (TBS, 20X), with 1% Tween 20 is used as a wash buffer and for sample preparation in general immunoassay applications, Tween 20 reduces nonspecific binding and protein: protein interactions.TBS is often used for diluting secondary antibodies or streptavidin-AP conjugate. TBS-Tween 20 is often used for washing steps.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TRIS-buffered saline (TBS, 20X), with 1% Tween 20 is used as a wash buffer and for sample preparation in general immunoassay applications, Tween 20 reduces nonspecific binding and protein: protein interactions.TBS is often used for diluting secondary antibodies or streptavidin-AP conjugate. TBS-Tween 20 is often used for washing steps.

Solubility
It is soluble in water.

Notes
250mM Tris, 27mM KCl, 1.37M NaCl, pH 7.4, with 1% Tween 20
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material TBS
Concentration 10X
Physical Form Liquid
Quantity 2 L

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.