missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TRIS-buffered saline (TBS, 10X) pH 7.4, for Western blot

Product Code. 15430277
Click to view available options
Quantity:
1 L
2 L
4 L
Unit Size:
1 L
2 L
4 L
This item is not returnable. View return policy

Product Code. 15430277

Brand: Thermo Scientific Chemicals J62938.K2

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

For Western blot washing | CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

For Western blot washing.TRIS-buffered saline (TBS, 10X) is used as a washing buffer for electrodes, alkaline phosphatase and peroxidase conjugates in dot blot assay. Further, it acts as a pH stabilizer, which enable washing without disruption of antibody-antigen binding interactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
For Western blot washing.TRIS-buffered saline (TBS, 10X) is used as a washing buffer for electrodes, alkaline phosphatase and peroxidase conjugates in dot blot assay. Further, it acts as a pH stabilizer, which enable washing without disruption of antibody-antigen binding interactions.

Solubility
Miscible with water.

Notes
500mM Tris, 1.5M NaCl, pH 7.4.Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Concentration 10X
pH 7.4
Physical Form Liquid
Quantity 1 L
Solubility Information Miscible with water.
Chemical Name or Material TRIS-buffered saline, for Western blot

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.