missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TRIS-buffered saline (TBS, 20X) pH 7.4

Product Code. 15413579
Change view
Click to view available options
Quantity:
1 L
2 L
Unit Size:
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15413579 1 L 1 L
15423579 2 L 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15413579 Supplier Thermo Scientific Chemicals Supplier No. J60877.K2
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

Tris Buffered Saline (TBS) is a pH stabilizing solution used for western blot and ELISA procedures. It is used as a convenient prepared buffer for use in various applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris Buffered Saline (TBS) is a pH stabilizing solution used for western blot and ELISA procedures. It is used as a convenient prepared buffer for use in various applications.

Solubility
It is soluble in water.

Notes
Contains: 0.5M Tris, 54mM KCl, and 2.74M NaCl, pH 7.4
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00132476
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Concentration 20X
pH 7.4
Physical Form Liquid
Quantity 1 L
Solubility Information It is soluble in water.
Chemical Name or Material TRIS-buffered saline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.