missing translation for 'onlineSavingsMsg'
Learn More

Tris(dibenzoylmethane)mono(phenanthroline)europium(III)

Product Code. 11389668
Change view
Click to view available options
Quantity:
0.1 g
0.5 g
Unit Size:
0.1g
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389668 0.1 g 0.1g
11399668 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389668 Supplier Thermo Scientific Alfa Aesar Supplier No. 044175.77
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. Rückgaberichtlinie anzeigen

As an organic red light emitter | CAS: 17904-83-5 | C57H44EuN2O6 | 1004.951 g/mol

As an organic red light emitter

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 17904-83-5
Molecular Formula C57H44EuN2O6
Molecular Weight (g/mol) 1004.951
MDL Number MFCD01321202
InChI Key DYKOLWWJTALFFU-RWBKAWJDSA-N
Synonym eu dbm 3 phen, tris dibenzoylmethane mono phenanthroline europium iii, tris dibenzoylmethane phenanthroline europium iii, tris dibenzoylmethane mono 1,10-phenanthroline europium lll, europium iii tris 1,3-diphenyl-1,3-propanedionato mono 1,10-phenanthroline, 2e-3-bis 1e-3-oxo-1,3-diphenylprop-1-en-1-yl oxy europio oxy-1,3-diphenylprop-2-en-1-one; phen
PubChem CID 14205791
IUPAC Name europium;(E)-3-hydroxy-1,3-diphenylprop-2-en-1-one;1,10-phenanthroline
SMILES C1=CC=C(C=C1)C(=CC(=O)C2=CC=CC=C2)O.C1=CC=C(C=C1)C(=CC(=O)C2=CC=CC=C2)O.C1=CC=C(C=C1)C(=CC(=O)C2=CC=CC=C2)O.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.[Eu]

Spezifikation

Melting Point 172°C to 173°C
Quantity 0.1 g
Solubility Information Soluble in dichloromethane, THF
Formula Weight 1001.93
Physical Form Powder
Chemical Name or Material Tris (dibenzoylmethane)mono (phenanthroline)europium (III)

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.