missing translation for 'onlineSavingsMsg'
Learn More

Tris(dibenzylideneacetone)dipalladium, TRC

Product Code. 30010814
Change view
Click to view available options
Quantity:
250 mg
10 g
25 g
Unit Size:
10g
250mg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010814 250 mg 250mg
30005781 25 g 25g
30006868 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010814 Supplier Toronto Research Chemicals Supplier No. TRCT876335250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51364-51-3
Molecular Formula 3 C17 H14 O . 2 Pd
Molecular Weight (g/mol) 915.72
Synonym (all-E)-Tris[μ-[(1,2-η:4,5-η)-1,5-diphenyl-1,4-pentadien-3-one]]di-palladium, (E,E)-1,5-Diphenyl-1,4-pentadien-3-one Palladium Complex, Tris(dibenzylideneacetonyl)bis-palladium, Tris[(1E,4E)-1,5-diphenyl-1,4-pentadien-3-one]dipalladium, Tris[μ-[(1,2-η:4,5-η)-(1E,4E)-1,5-diphenyl-1,4-pentadien-3-one]]dipalladium, tris[μ-[(1,2-η:4,5-η)-(1E,4E)-1,5-diphenyl-1,4-pentadien-3-one]]dipalladium [Pd2(dba)3]
IUPAC Name (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
SMILES [Pd].[Pd].O=C(\C=C\c1ccccc1)\C=C\c2ccccc2.O=C(\C=C\c3ccccc3)\C=C\c4ccccc4.O=C(\C=C\c5ccccc5)\C=C\c6ccccc6

Specifications

Quantity 250 mg
InChI Formula InChI=1S/3C17H14O.2Pd/c3*18-17(13-11-15-7-3-1-4-8-15)14-12-16-9-5-2-6-10-16;;/h3*1-14H;;/b3*13-11+,14-12+;;
Formula Weight 914.1204
Percent Purity >95%
Chemical Name or Material Tris(dibenzylideneacetone)dipalladium
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.