missing translation for 'onlineSavingsMsg'
Learn More

Tris-EDTA Buffer Solutions, R&D Systems™

Product Code. 18064685 Shop All Bio Techne Products
Change view
Click to view available options
pH:
8
9
Unit Size:
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Unit Size
18064685 9 500mL
18010384 8 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18064685 Supplier Novus Biologicals Supplier No. NB90062085
  • ONLINE EXCLUSIVE PRICE
    542.25 €
    471.45 € / 500mL
    Save 70.80 €
    13% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. For recovery of antigens masked by fixation in cross-linking fixatives, such as formalin

  • 10X EDTA buffer for Immunohistochemistry
  • Dilute one part buffer with nine parts distilled water
  • Unit size: 500mL

Recommended Storage

Store at room temperature

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Retrieval of antigens masked by fixation in cross linking fixatives such as formalin
pH 9
Product Line Novus
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.