missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris-Glycine Buffer, 10X Powder, Electrophoresis, Fisher BioReagents
Description
- Used to make a 10X concentrate solution that can be diluted for use as a transfer buffer for Western blotting and gel electrophoresis
- DNase, RNase, and protease free powder that is pre-mixed for your convenience to make 1L of 10X solution
- Eliminates the need to separately weigh ingredients - simply dissolve powder in pure water
Specifications
Specifications
| Chemical Name or Material | Tris-Glycine |
| Name Note | 10X Powder |
| CAS | 77-86-1, 56-40-6 |
| Color | White |
| pH | 8.3 |
| Physical Form | Powder or Solid |
| Quantity | 1 Pk. |
| DNase | DNase free |
| Packaging | Foil Pack |
| Protease | Protease free |
| Show More |
Safety and Handling
Product Identifier
- Tris Glycine, 10X Powder
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?