missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TRIS-glycine-SDS running buffer (10X), pH 8.5

Product Code. 15493669
Change view
Click to view available options
Quantity:
1 L
2 L
4 L
Unit Size:
1 L
2 L
4 L
Product Code. Quantity unitSize
15493669 1 L 1 L
15403679 2 L 2 L
15413679 4 L 4 L
3 options
This item is not returnable. View return policy

Product Code. 15493669

Brand: Thermo Scientific Chemicals J61006.K2

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TRIS-glycine-SDS running buffer (10X), suitable for protein gel analysis, is used in several research areas, including biochemistry, molecular biology, cell biology, and immunology.

Tris-Glycine SDS Running Buffer (10X) is used as the electrophoresis buffer during the stacking and resolve process of sodium dodecyl sulfate - polyacrylamide gel electrophoresis (SDS-PAGE). It is ideally suited for protein gel analysis in biochemical, molecular biological, cell biological,and immunological research studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• TRIS-glycine-SDS running buffer (10X), is the most commonly used buffer for sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) of proteins

Application

• This solution is the most commonly used buffer for protein gel analysis in several research studies
• It is also used to block membranes with 5% non-fat dry milk for western blotting
• TRIS-glycine-SDS running buffer is commonly used as both anode buffer and cathode buffer
• The use of SDS in the buffer helps keep the proteins linear. Glycine is an amino acid whose charge state plays an important role in the stacking gel

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material TRIS-glycine-SDS running buffer (10X)
pH 8.5
Physical Form Liquid
Quantity 1 L
Packaging Plastic bottle
Solubility Information It is soluble in water.
What are the concentrations of Tris base, glycine and SDS in the TRIS-glycine-SDS running buffer (10X), pH 8.3?

The TRIS-glycine-SDS running buffer (10X), pH 8.3 contains 0.25 M Tris base, 1.92 M glycine, and 1% SDS.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.