missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris Hydrochloride, 1M Solution (pH 7.0/Mol. Biol.), Fisher BioReagents™
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Poly Bottle
Unit Size:
100mL
500mL
Description
Tris Hydrochloride, 1M Solution (pH 7.0/Mol. Biol.)
Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. These 1M solutions of Tris Hydrochloride at pH 7.0 provide convenient and highly purified solutions.
Specifications
Specifications
| Color | Undesignated |
| Concentration | 0.96 to 1.04 M |
| pH | 10 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| DNase | DNase free |
| Filtered Through | Filtered through a 0.2-micron filter. |
| Identification | Pass Test |
| Packaging | Poly Bottle |
| Protease | Protease free |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction