missing translation for 'onlineSavingsMsg'
Learn More

TRIS hydrochloride, >99%, Cell culture reagent grade, MP Biomedicals™

Product Code. 11474922 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g

Product Code. 11474922

Brand: MP Biomedicals 0219455880

Please to purchase this item. Need a web account? Register with us today!

Excellent biochemical and biological buffer for all applications where ultra high purity is required. The pH values of all buffers are temperature and concentration dependent

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
MDL Number MFCD00012590
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
Synonym TRIS HCl, Tris(hydroxymethyl)aminomethane hydrochloride
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Color White
Physical Form Crystalline Powder
Quantity 100 g
Formula Weight 157.6
Grade Cell Culture
Chemical Name or Material TRIS hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.