missing translation for 'onlineSavingsMsg'
Learn More

Tris(hydroxymethyl)aminomethane, 99.8%, ACS reagent

Product Code. 10424521 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10424521 500 g Plastic bottle 500g
10731072 100 g Plastic bottle 100g
11985281 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10424521 Supplier Thermo Scientific Acros Supplier No. 424575000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym 2-Amino-2-(hydroxymethyl)-1, 3-propanediol, TRIS, Tris buffer, Tromethamine, Tromethane
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Absorbance 0.2 max. at 290nm, (40% aq. soln.) (1 cm cell), (40% aq. soln.) (1 cm cell) at 0nm
Absorbance Conditions (40% aq. soln.) (1 cm cell)
Melting Point >168.5°C
Boiling Point 219.0°C to 220.0°C (10.0mmHg)
Color White
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 500 g
Assay Percent Range 99.8%
Infrared Spectrum Authentic
Linear Formula (HOCH2)3CNH2
Packaging Plastic bottle
Beilstein 04, 303
Merck Index 15, 9951
Formula Weight 121.14
Percent Purity 99.8%
Grade ACS Reagent
Chemical Name or Material Tris(hydroxymethyl)aminomethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.