missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane hydrochloride, 1M soln., pH 7,4, RNase free

Product Code. 15415805
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15415805 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15415805 Supplier Thermo Scientific Chemicals Supplier No. J62848.AK
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol

Tris Buffered Saline (TBS) is a pH stabilizing solution used for western blot and ELISA procedures. It is used as a convenient prepared buffer for use in various applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
CAS 1185-53-1
Concentration 1M soln.
Molecular Formula C4H12ClNO3
pH 7.4
MDL Number MFCD00012590
Quantity 250 mL
Physical Form Liquid
Synonym tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride
Solubility Information It is soluble in water.
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
SMILES C(C(CO)(CO)N)O.Cl
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
Molecular Weight (g/mol) 157.594
PubChem CID 93573
Chemical Name or Material Tris(hydroxymethyl)aminomethane hydrochloride, RNAse free
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.