missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane hydrochloride, 1M stock soln, pH 8.0

Product Code. 15430317
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15430317 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15430317 Supplier Thermo Scientific Chemicals Supplier No. J63636.K2
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol

Tris, Hydrochloride is commonly used as a buffer component for phenol extractions of DNA or RNA and a buffering component of separating and stacking gels in the characterization of protein products by SDS-PAGE. Tris, Hydrochloride has also been used with urea as a method for antigen retrieval in immunohistochemistry. In smooth muscles Tris, Hydrochloride has been observed to inhibit motor responses to adrenergic motor nerve stimulation. Also uses include pH control in buffering systems for electrophoresis applications. Tris has been used as a starting material for polymers, oxazolones (with carboxylic acids) and oxazolidines (with aldehydes).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
CAS 1185-53-1
Molecular Formula C4H12ClNO3
MDL Number MFCD00012590
Concentration 1M stock soln.
Synonym tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride
Solubility Information Fully miscible with water.
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
SMILES C(C(CO)(CO)N)O.Cl
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
Molecular Weight (g/mol) 157.594
PubChem CID 93573
pH 8.0
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Tris(hydroxymethyl)aminomethane hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.