missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane hydrochloride, Molecular Biology Grade, 99+%

Product Code. 15488679
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488679 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15488679 Supplier Thermo Scientific Chemicals Supplier No. J67233.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
  • Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications
TRUSTED_SUSTAINABILITY

Specifications

CAS 1185-53-1
Melting Point ∽152°C (decomposition)
Color White
pH 3.5 to 5.0
Physical Form Powder
Quantity 100 g
Molecular Formula C4H12ClNO3
MDL Number MFCD00012590
Beilstein 3675235
Synonym TRIS hydrochloride; 2-Amino-2-hydroxymethyl-1,3-propanediol hydrochloride
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
SMILES C(C(CO)(CO)N)O.Cl
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
Molecular Weight (g/mol) 157.594
PubChem CID 93573
Formula Weight 157.6
Percent Purity ≥99%
Grade Molecular Biology
Chemical Name or Material Tris(hydroxymethyl)aminomethane hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.