missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(pentafluorophenyl)borane, 95%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Spezifikation
| Melting Point | 126°C to 131°C |
| Elemental Analysis | 94% min. |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 94% min |
| Packaging | Glass bottle |
| Linear Formula | (C6F5)3B |
| Quantity | 1 g |
| Merck Index | 15, 9931 |
| Formula Weight | 511.99 |
| Percent Purity | 95% |
| Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- Tris(pentafluorophenyl)borane
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur