missing translation for 'onlineSavingsMsg'
Learn More

Trithiocyanuric acid, 95%

Product Code. 11479376
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479376 25 g 25g
11469376 5 g 5g
11489376 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479376 Supplier Thermo Scientific Alfa Aesar Supplier No. A19080.14
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 638-16-4 | C3H3N3S3 | 177.258 g/mol

Reaction of BunLi with trithiocyanuric acid (LH3) in toluene/HMPA yielded a mono-lithiated species, [(LH2Li)·HMPA2]. Metal complexes of trithiocyanuric acid is used for the elucidation of coordination polymer stoichiometry via thermometric titrimetry:.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reaction of BunLi with trithiocyanuric acid (LH3) in toluene/HMPA yielded a mono-lithiated species, [(LH2Li)·HMPA2]. Metal complexes of trithiocyanuric acid is used for the elucidation of coordination polymer stoichiometry via thermometric titrimetry:.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-16-4
Molecular Formula C3H3N3S3
Molecular Weight (g/mol) 177.258
MDL Number MFCD00006052
InChI Key WZRRRFSJFQTGGB-UHFFFAOYSA-N
Synonym trithiocyanuric acid, thiocyanuric acid, 1,3,5-triazine-2,4,6-trithiol, trismercaptotriazine, trimercaptocyanuric acid, cyanuric acid, trithio, usaf th-3, 1,3,5-triazine-2,4,6 1h,3h,5h-trithione, s-triazine-2,4,6-trithiol, 2,4,6-triazinetrithiol
PubChem CID 1268121
IUPAC Name 1,3,5-triazinane-2,4,6-trithione
SMILES C1(=S)NC(=S)NC(=S)N1

Specifications

Melting Point >300°C
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 177.27
Percent Purity 95%
Chemical Name or Material Trithiocyanuric acid
compliance-icons
Product Identifier
  • Trithiocyanuric acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Trithiocyanuric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.