missing translation for 'onlineSavingsMsg'
Learn More

Triton X-100, TRC

Product Code. 30453635
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453635 10 g 10g
30452837 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30453635 Supplier Toronto Research Chemicals Supplier No. TRCT88680010G
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C14H22O.(OCH2CH2)n
Molecular Weight (g/mol) 250.38
Synonym Polyethyleneglycols Mono[p-(1,1,3,3-tetramethylbutyl)phenyl] Ether, p-(1,1,3,3-Tetramethylbutyl)phenol Monoether with Polyethylene Glycol, (p-t-Octylphenoxy)polyethoxyethanol, Anapoe X 100, Anapoe X 114, Antarox A 200, Coulter Dispersant IA, Criton X 100, Hydrol SW, Iconol OP, Intra-Cell, Koromex II, Lutensol OP 10, NOP 90, Nonidel P 40, OPE 30, Octoxynol 5, Ortho-Gynol, Photo-Flow 200, Poly(oxyethylene) p-tert-Octylphenyl Ether, Polyethylene Gllycol Mono(4-octylphenyl) Ether, Polyethylene Glycol Mono(4-tert-octylphenyl) Ether, Polyoxyethylene Glycol-p-tert-octylphenyl Ether, Polyoxyethylene Mono(octylphenyl) Ether, Preceptin, Surfac OP 30, Surfac OP 5, Surfonic OP 120, TOP 10, TX 100, TX 102, TX 305, TX 35, TX 405, Texofor FP 300, Octoxynol 9, Octylphenoxy poly(ethyleneoxy)ethanol, branched
IUPAC Name 14-(4-(2,4,4-trimethylpentan-2-yl)phenoxy)-3,6,9,12-tetraoxatetradecan-1-ol
SMILES CC(C(C=C1)=CC=C1OCCOCCOCCOCCOCCO)(C)CC(C)(C)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C24H42O6/c1-23(2,3)20-24(4,5)21-6-8-22(9-7-21)30-19-18-29-17-16-28-15-14-27-13-12-26-11-10-25/h6-9,25H,10-20H2,1-5H3
Formula Weight 250.1933
Chemical Name or Material Octoxinol 10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.