missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tritosulfuron metabolite BH635-4 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Tritosulfuron Metabolite BH635-4 |
| CAS | 1418095-29-0 |
| Analyte or Component Names | Tritosulfuron Metabolite BH635-4 |
| Shipping Condition | -18°C |
| Molecular Formula | C10 H10 F3 N5 O4 S |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Benzenesulfonamide, N-[[[[(aminocarbonyl)amino]iminomethyl]amino]carbonyl]-2-(trifluoromethyl)- |
| InChI Formula | InChI=1S/C10H10F3N5O4S/c11-10(12,13)5-3-1-2-4-6(5)23(21,22)18-9(20)17-7(14)16-8(15)19/h1-4H,(H6,14,15,16,17,18,19,20) |
| SMILES | NC(=O)NC(=N)NC(=O)NS(=O)(=O)c1ccccc1C(F)(F)F |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?