missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRK fused gene Recombinant Rabbit Monoclonal Antibody (24GB2115)

Rabbit Recombinant Monoclonal Antibody
198.00 € - 564.00 €
Specifications
| Antigen | TRK fused gene |
|---|---|
| Clone | 24GB2115 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275648
![]() |
Invitrogen™
MA557800 |
100 μL |
564.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275199
|
Invitrogen™
MA557801 |
20 μL |
198.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRK fused gene Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
TFG protein, a recently identified novel protein, is a human homolog of mouse TFG protein. It contains an N-terminal coiled-coil structure, a SPYGQ-rich region followed by octicosapeptide repeats. Research shows that TFG can modulate SHP-1 activity and is a novel member of the NF-kB pathway. The physiological functions of this protein remain largely elusive, but TFG is thought to function by associating with other proteins, including TANK and NEMO. TFG was first identified as a partner of NTRK1 and is involved in generating the thyroid TRK-T3 oncogene, as well as oncogenic rearrangements with ALK in anaplastic lymphoma and NOR1 in mixoid chondrosarcoma.Specifications
| TRK fused gene | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| TFG | |
| AI173908; HMSNP; protein TFG; SPG57; TF6; TFG; Trk-fused gene; TRK-fused gene protein; TRKT3; TRKT3 oncogene | |
| TFG | |
| Primary | |
| Affinity chromatography |
| 24GB2115 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q92734 | |
| 10342, 21787, 360709 | |
| A synthesized peptide derived from human TRK fused gene (aa 362-400). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title