missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TrkC Antibody Pair (HRP), Novus Biologicals™
Mouse Monoclonal Antibody
438.00 €
Specifications
| Antigen | TrkC |
|---|---|
| Dilution | Sandwich ELISA |
| Classification | Monoclonal |
| Conjugate | HRP |
| Host Species | Mouse |
Description
TrkC Monoclonal specifically detects TrkC in Human samples. It is validated for ELISA, Sandwich ELISA.Specifications
| TrkC | |
| Monoclonal | |
| Mouse | |
| EC 2.7.10, EC 2.7.10.1, ETS related protein-neurotrophic receptor tyrosine kinase fusion protein, ETV6-NTRK3 fusion, gp145(trkC), GP145-TrkC, Neurotrophic tyrosine kinase receptor type 3, neurotrophic tyrosine kinase, receptor, type 3, NT 3 receptor, NT-3 growth factor receptor, Trk-C, TrkC tyrosine kinase, TRKCneurotrophin 3 receptor, tyrosine kinase receptor C | |
| NTRK3 | |
| This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TrkC. |
| Sandwich ELISA | |
| HRP | |
| Cancer, Phospho Specific, Protein Kinase | |
| 4916 | |
| Storage of components varies. See protocol for specific instructions. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title