missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRMT11 Polyclonal Antibody

Product Code. 15515932
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15515932 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15515932 Supplier Invitrogen™ Supplier No. PA543190

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: IKIHTFNKTL TQEEKIKRID ALEFLPFEGK VNLKKPQHVF SVLEDYGLDP Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

TRMT11 belongs to the methyltransferase superfamily. It is a catalytic subunit of an S-adenosyl-L-methionine-dependent tRNA methyltransferase complex that mediates the methylation of the guanosine nucleotide at position 10 (m2G10) in tRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRMT11
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TRMT11
Gene Accession No. Q7Z4G4
Gene Alias 2410075D05Rik; 3110045I18Rik; AW213713; C6orf75; MDS024; putative RNA methylase MDS024; TRM11; Trmt11; TRMT11-1; tRNA (guanine(10)-N2)-methyltransferase homolog; tRNA guanosine-2'-O-methyltransferase TRM11 homolog; tRNA guanosine-2'-O-methyltransferase TRM11-like protein-like protein; tRNA methyltransferase 11; tRNA methyltransferase 11 homolog; tRNA methyltransferase 11 homolog (S. cerevisiae)
Gene Symbols TRMT11
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TRMT11 (aa 110-159).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60487
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.