missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRNT1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94361-0.1ml
This item is not returnable.
View return policy
Description
TRNT1 Polyclonal antibody specifically detects TRNT1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| TRNT1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CCA1, CCA-adding, CGI-47, EC 2.7.7.72, mitochondrial CCA-adding tRNA-nucleotidyltransferase, mt CCA-adding enzyme, mt tRNA adenylyltransferase, mt tRNA CCA-diphosphorylase, mt tRNA CCA-pyrophosphorylase, MtCCA, tRNA nucleotidyl transferase, CCA-adding, 1, tRNA-nucleotidyltransferase 1, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 135-434 of human TRNT1 (NP_886552.2). HAEVEFTTDWQKDAERRDLTINSMFLGFDGTLFDYFNGYEDLKNKKVRFVGHAKQRIQEDYLRILRYFRFYGRIVDKPGDHDPETLEAIAENAKGLAGISGERIWVELKKILVGNHVNHLIHLIYDLDVAPYIGLPANASLEEFDKVSKNVDGFSPKPVTLLASLFKVQDDVTKLDLRLKIAKEEKNLGLFIVKNRKDLIKATDSSDPLKPYQDFIIDSREPDATTRVCELLKYQGEHCLLKEMQQWSIPPFPVSGHDIRKVGISSGKEIGALLQQLREQWKKSGYQMEKDELLSYIKKT | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51095 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction