missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TROP-2 Antibody (rTACSTD2/9326), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23715-20ug
This item is not returnable.
View return policy
Description
TROP-2 Monoclonal antibody specifically detects TROP-2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TROP-2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Cell surface glycoprotein Trop-2, EGP1, EGP-1, epithelial glycoprotein-1, GA733-1GP50, gastrointestinal tumor-associated antigen GA7331, M1S1cell surface glycoprotein TROP2, Membrane component chromosome 1 surface marker 1, membrane component, chromosome 1, surface marker 1, Pancreatic carcinoma marker protein GA733-1, pancreatic carcinoma marker protein GA7331,40kD glycoprotein, identified by monoclonal GA733, TROP2GA7331, tumor-associated calcium signal transducer 2 | |
| Recombinant fragment of human TROP-2 protein (around aa 31-274) (exact sequence is proprietary) | |
| 20 μg | |
| Signal Transduction | |
| 4070 | |
| Store at 4°C. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| rTACSTD2/9326 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction