missing translation for 'onlineSavingsMsg'
Learn More

Tropicamide, MedChemExpress

Product Code. 16482096
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482096 500 mg 500mg
16408236 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16482096 Supplier Medchem Express Supplier No. HYB0321
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tropicamide (Ro 1-7683) is a selective M4 muscarinic acetylcholine receptor antagonist. Tropicamide produces short acting mydriasis (dilation of the pupil) and cycloplegia when applied as eye drops.

Tropicamide (Ro 1-7683) is a selective M4 muscarinic acetylcholine receptor antagonist. Tropicamide produces short acting mydriasis (dilation of the pupil) and cycloplegia when applied as eye drops.

Specifications

Chemical Name or Material Tropicamide
Physical Form Solid
CAS 1508-75-4
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C17H20N2O2
For Use With (Application) Neuroscience-Neuromodulation
Synonym Ro 1-7683
Solubility Information DMSO : 25 mg/mL (87.92 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.36 mM; Need ultrasonic)
SMILES O=C(N(CC)CC1=CC=NC=C1)C(C2=CC=CC=C2)CO
Molecular Weight (g/mol) 284.35
Formula Weight 284.35
Percent Purity 99.3%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.