missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Tropodithietic acid Broad spectrum antibiotic

Product Code. 18297533 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18297533

Brand: Novus Biologicals™ NBP262760

This item is not returnable. View return policy

Broad spectrum antibiotic

Molecular Weight: 212.3
PubChem ID: 44632924

Application Notes: May be dissolved in DMSO (5 mg/mL).

Specifications

CAS 750590-18-2
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
For Use With (Application) Orange Powder. May be Dissolved in DMSO (5 mg/mL).
Purity >98% (HPLC); NMR (conforms)
Quantity 1 mg
Source Synthetic
Target Tropodithietic acid Broad spectrum antibiotic
Form Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.