missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tropomyosin-1 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NBP2-29883-0.025ML
This item is not returnable.
View return policy
Description
Tropomyosin-1 Polyclonal specifically detects Tropomyosin-1 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Tropomyosin-1 | |
| Polyclonal | |
| Western Blot 1:200-1:1000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500 | |
| P09493 | |
| TPM1 | |
| Synthetic peptide corresponding to the 9d exon from the alpha gene of tropomyosin. | |
| 0.025 mL | |
| Cytoskeleton Markers | |
| 7168 | |
| Mouse, Rat | |
| IgG |
| Immunocytochemistry, Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS with No Preservative | |
| alpha tropomyosin, alpha-tropomyosin, C15orf13, cardiomyopathy, hypertrophic 3, chromosome 15 open reading frame 13, CMD1Y, CMH3, HTM-alpha, sarcomeric tropomyosin kappa, TMSA, tropomyosin 1 (alpha), tropomyosin 1 (alpha) isoform 1, tropomyosin 1 (alpha) isoform 2, tropomyosin 1 (alpha) isoform 3, tropomyosin 1 (alpha) isoform 4, tropomyosin 1 (alpha) isoform 5, tropomyosin 1 (alpha) isoform 6, tropomyosin 1 (alpha) isoform 7, tropomyosin alpha-1 chain, Tropomyosin-1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Recognizes Tropomyosin 1, 2, 3, 5a, 5b, 6. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur