Learn More
TRPA1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 41 publication
Brand:  Novus Biologicals NB110-40763
 View more versions of this product
Product Code. 18728053
  • 623.00 € / 0.10mL
Description
TRPA1 Polyclonal specifically detects TRPA1 in Human, Mouse, Rat, Guinea Pig, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TRPA1
ELISA
A synthetic peptide made to a region within the N-terminus (residues 1-100) of the human TRPA1 protein. [Swiss-Prot# O75762]
0.1 mL