missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPA1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 41 publication
Brand: Novus Biologicals NB110-40763SS
This item is not returnable.
View return policy
Description
TRPA1 Polyclonal specifically detects TRPA1 in Human, Mouse, Rat, Guinea Pig, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| TRPA1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| ANKTM1ankyrin-like with transmembrane domains 1, Ankyrin-like with transmembrane domains protein 1, Transformation-sensitive protein p120, transient receptor potential cation channel subfamily A member 1, transient receptor potential cation channel, subfamily A, member 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Additional modified form of TRPA1 can also be detected. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 2 ug/ml, Flow Cytometry reported in scientific literature (PMID 28990934), ELISA reported in scientific literature (PMID 29803505), Immunohistochemistry 0.5-1.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:100-1:250, Immunohistochemistry-Frozen reported in scientific literature | |
| O75762 | |
| TRPA1 | |
| A synthetic peptide made to a region within the N-terminus (residues 1-100) of the human TRPA1 protein. [Swiss-Prot# O75762] | |
| 0.025 mL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Sensory Systems | |
| 8989 | |
| Human, Mouse, Rat, Guinea Pig | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction