missing translation for 'onlineSavingsMsg'
Learn More

TRPM1 Polyclonal Antibody, Invitrogen™

Product Code. 11525073
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11525073 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11525073 Supplier Invitrogen Supplier No. OSR00187W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

In IHC (P) applications, heating the tissue sections above 85°C is not recommended. Reconstitute with 100 μL of sterile water.

The protein encoded by this gene is similar to the transient receptor potential (Trp) calcium channel family members. The expression of this protein is inversely correlated with melanoma aggressiveness, suggesting that it suppresses melanoma metastasis.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen TRPM1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene TRPM1
Gene Accession No. Q7Z4N2
Gene Alias Transient receptor potential cation channel subfamily M member 1, TRPM1, MLSN1, LTRPC1, melastatin-1, melastatin 1
Gene Symbols TRPM1
Host Species Rabbit
Immunogen A synthetic peptide from aa region 750-800 of human TRPM1 conjugated to an immunogenic carrier protein was used as the antigen
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4308
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.