Learn More
TRPM2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21842-1-AP-20UL
This item is not returnable.
View return policy
Description
The TRPM2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TRPM2. This antibody recognizes human, mouse antigen. The TRPM2 antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
| TRPM2 | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Immunofluorescence, Immunohistochemistry, ELISA | |
| EREG1, KNP3, LTrpC 2, LTRPC2, NUDT9H, NUDT9L1, TRPC7, TRPM2 | |
| 20 μL | |
| 7226 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |