Learn More
Invitrogen™ TRPM7 Recombinant Rabbit Monoclonal Antibody (24GB2250)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557806
 View more versions of this product
Product Code. 30276102
  • 564.00 € / 100µL
Description
TRPM7 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
TRPCs, mammalian homologs of the Drosophila transient receptor potential (trp) protein, are ion channels that are thought to mediate capacitative calcium entry into the cell. TRP-PLIK is a protein that is both an ion channel and a kinase. As a channel, it conducts calcium and monovalent cations to depolarize cells and increase intracellular calcium. As a kinase, it is capable of phosphorylating itself and other substrates. The kinase activity is necessary for channel function, as shown by its dependence on intracellular ATP and by the kinase mutants.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TRPM7
Recombinant Monoclonal
Unconjugated
Trpm7
2310022G15Rik; 4833414K03Rik; 5033407O22Rik; ALSPDC; CHAK; CHAK1; Channel kinase 1; Channel-kinase 1; FLJ20117; long transient receptor potential channel 7; long transient receptor potential channel 7 {ECO:0000250; Ltpr7; LTRPC ion channel family member 7; LTRPC7; LTrpC-7; LTrpC7 {ECO:0000250; PubMed:11161216}; RGD:620053}; tr; transient receptor potential cation channel subfamily M member 7; transient receptor potential cation channel subfamily M member 7 {ECO:0000312; transient receptor potential cation channel, subfamily M, member 7; transient receptor potential M7; transient receptor potential-phospholipase C-interacting kinase; transient receptor potential-phospholipase C-interacting kinase {ECO:0000303; transient receptor potential-related protein, ChaK; TRMP7; TRP PLIK; Trpm7; trpm7 {ECO:0000312; TRPPLIK; Trp-plik; TRP-PLIK {ECO:0000303; UniProtKB:Q96QT4}
Rabbit
Affinity chromatography
RUO
54822
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2250
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q96QT4
Trpm7
A synthesized peptide derived from human TRPM7 (aa 1800-1865).