missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPM8 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
190.00 € - 531.00 €
Specifications
| Antigen | TRPM8 |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:300, KnockDown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18799623
|
Novus Biologicals
NBP1-97311SS |
0.025 mL |
190.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18750424
|
Novus Biologicals
NBP1-97311 |
0.1 mL |
531.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRPM8 Polyclonal specifically detects TRPM8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| TRPM8 | |
| Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:300, KnockDown Validated | |
| Polyclonal | |
| Rabbit | |
| Cancer, Tumor Suppressors | |
| PBS (pH 7.4) with 0.05% Sodium Azide | |
| Long transient receptor potential channel 6, LTrpC6, LTrpC-6, MGC2849, short form of the TRPM8 cationic channel, transient receptor potential cation channel, subfamily M, member 8, Transient receptor potential p8, transient receptor potential subfamily M member 8, trp-p8, TRPP8transient receptor potential cation channel subfamily M member 8 | |
| TRPM8 | |
| IgG | |
| Affinity Purified |
| 1.0 mg/ml | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| Q7Z2W7, Q8R4D5, Q8R455 | |
| 79054 | |
| A synthetic peptide made to an internal portion of the human TRPM8 protein (between residues 250-300) [UniProt# Q7Z2W7] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title