missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPV5 Antibody (SR2284), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22511-100ul
This item is not returnable.
View return policy
Description
TRPV5 Monoclonal antibody specifically detects TRPV5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| TRPV5 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR2284 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Calcium transport protein 2, calcium transporter 2, ECaC, ECaC1, ECAC1CaT2, Epithelial calcium channel 1CAT2, Osm-9-like TRP channel 3, OTRPC3, transient receptor potential cation channel subfamily V member 5, transient receptor potential cation channel, subfamily V, member 5, TrpV5 | |
| A synthesized peptide derived from human TRPV5 (Uniprot #: Q9NQA5) | |
| 100 μg | |
| Cancer, Endocrinology, Signal Transduction | |
| 56302 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction