missing translation for 'onlineSavingsMsg'
Learn More

3'-Nitroacetanilide, 98+%

Artikelnummer. 11499557
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
5 g
25 g
100 g
Pakningsstørrelse:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
11499557 5 g 5g
11409567 25 g 25g
11419567 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11499557 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. B22062.06
 Restricted Item
In Stock
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format
Denne vare kan ikke returneres. Se returpolicy

CAS: 122-28-1 | C8H8N2O3 | 180.163 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 122-28-1
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.163
MDL Number MFCD00017015
InChI Key KFTYNYHJHKCRKU-UHFFFAOYSA-N
Synonym n-3-nitrophenyl acetamide, 3'-nitroacetanilide, m-nitroacetanilide, 3-nitroacetanilide, acetamide, n-3-nitrophenyl, n-acetyl-m-nitroaniline, 3-nitro-n-acetylaniline, acetanilide, 3'-nitro, unii-qgh8s22nbp, qgh8s22nbp
PubChem CID 31206
IUPAC Name N-(3-nitrophenyl)acetamide
SMILES CC(=O)NC1=CC(=CC=C1)[N+](=O)[O-]

Tekniske data

Melting Point 151°C to 153°C
Quantity 5 g
Beilstein 2211961
Merck Index 14,6581
Formula Weight 180.16
Percent Purity ≥98%
Chemical Name or Material 3'-Nitroacetanilide

RUO – Research Use Only

Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.