missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptase alpha/beta 1 Antibody (TPSAB1/1961), Novus Biologicals™
Mouse Monoclonal Antibody
277.00 € - 474.00 €
Specifications
| Antigen | Tryptase alpha/beta 1 |
|---|---|
| Clone | TPSAB1/1961 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18641758
|
Novus Biologicals
NBP2-75751-20ug |
20 μg |
277.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18649077
|
Novus Biologicals
NBP2-75751-100ug |
100 μg |
474.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tryptase alpha/beta 1 Monoclonal antibody specifically detects Tryptase alpha/beta 1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Protein ArraySpecifications
| Tryptase alpha/beta 1 | |
| 0.2 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| Unconjugated | |
| Mouse | |
| Immunology, metabolism | |
| 10 mM PBS with 0.05% BSA | |
| EC:3.4.21.59, epididymis secretory sperm binding protein, mast cell alpha II tryptase, mast cell beta I tryptase, mast cell tryptase, TPS1, TPS2, TPSB1, TPSB2, tryptase alpha II, Tryptase alpha-1, tryptase beta I, tryptase beta-1, Tryptase beta-2, Tryptase I, tryptase II, tryptase-1, tryptase-2, tryptase-I, tryptase-III | |
| Recombinant human Tryptase alpha/TPS1 protein fragment (around aa 115-233) (Exact sequence is proprietary) (Uniprot: Q15661) | |
| Primary | |
| Store at 4°C. |
| TPSAB1/1961 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q15661 | |
| 7177 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title