missing translation for 'onlineSavingsMsg'
Learn More

BD Trypticase™ Soy Agar

Product Code. 11860842
Change view
Click to view available options
Product Type:
Agar Plate
Unit Size:
Pack of 120
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11860842 Agar Plate Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11860842 Supplier BD Supplier No. 254086
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pre-plated, ready-to-use aseptic media supports the growth of a variety of bacteria and fungi. BD Trypticase™ Soy Agar is supplied in sterile 90mm Stacker™ plates, packaged with 10 plates per sleeve.

BD Trypticase™ Soy Agar is a general purpose medium which supports the growth of nonfastidious and moderately fastidious microorganisms.
  • Formula (per 1L purified water):
    • 15g pancreatic digest of Casein
    • 5g pancreatic digest of Soybean
    • 5g Sodium Chloride (NaCl)
    • 15g Agar
  • Casein and soy peptones supply organic nitrogen, amino acids and longer-chained peptides
  • Sodium chloride maintains the osmotic equilibrium
  • Not recommended for the primary isolation of microorganisms from clinical specimens
  • May be used for subcultivating bacterial strains isolated from clinical specimens to obtain pure cultures
  • May also be used for further differentiation and identification
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store plates in the dark at 2-8°C, in their original sleeve wrapping until just prior to use
Product Type Agar Plate
Includes 90mm Stacker™ plates
No. per Pack 10 Plates/Sleeve
Packaging Type Sleeve wrapping
Quantity 120 Plates
Diameter (Metric) 90 mm Plates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.