missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tryptone Soy Agar with Disinhibitor, Contact Plate

Product Code. 11934172
Change view
Click to view available options
Description:
Tryptone Soy Agar with Disinhibitor Contact Plate
Format:
55mm Contact Plate
Quantity:
2 x 10 plates
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Format Quantity Unit Size
11934172 Tryptone Soy Agar with Disinhibitor Contact Plate 55mm Contact Plate 2 x 10 plates Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11934172 Supplier Thermo Scientific™ Supplier No. PO5024C
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Effortlessly sample surfaces after cleaning and disinfection with a TSA Contact Plate with disinhibitors to neutralize surface disinfectants and aid in microorganism recovery.

Effortlessly sample surfaces after cleaning and disinfection using Thermo Scientific™ Tryptone Soy Agar with Disinhibitor Contact Plates, containing Lecithin and polysorbate 80, to neutralize surface disinfectants and aid in microorganism recovery.

Prevent agar from falling out of the dish with Thermo Scientific Contact Plates, which feature a dome-shaped bottom. GripFit™ technology makes plates easy-to-handle and ensures the lid remains in place during handling, while being easily removed during environmental monitoring. An inner layer on the base of the plate is sub-divided into grids to allow easy calculation of colony forming units (CFU) from surface tests, enabling labs to report whether contaminant levels fall within pre-defined acceptable limits. Easy stacking provides more stability during handling and transport, and ventilation is reduced to prevent plates from drying out.

Our contact plates are manufactured using a specialized, automated procedure to ensure a smooth, evenly distributed layer of agar on each plate. Surface sampling is achieved by gently rolling the domed surface of the agar onto the area to be tested. If any microorganisms that can cause disease or infection are present, they will grow to form easily visible colonies on the surface of the agar. This offers a reliable and effective way of determining the microbial cleanliness of surface areas and help control invisible dangers associated with microbial contamination. Clearly visible growth is available within three days for bacteria and within five days for fungi.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Tested in accordance with BP/EP/JP/USP.
Color Ivory, transparent
Content And Storage 2 to 25°C
Description Tryptone Soy Agar with Disinhibitor Contact Plate
Format 55mm Contact Plate
Manufacturing Origin Germany
Product Type Agar Plate
Fill Weight 13.5g ± 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 2 x 10 plates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.